pVectOZ™ Transfection Plasmids, OZ Biosciences
Supplier: OZ Biosciences
PL00020 PL00030 PL00040 PL00050 PL00150 PL00010 PL00110 PL00120 PL00130 PL00140
103258-172EA 405.86 USD
103258-172 103258-174 103258-176 103258-178 103258-180 103258-330 103258-420 103258-422 103258-424 103258-426
pVectOZ™ Transfection Plasmids, OZ Biosciences
Transfection Reagents
pVectOZ Transfection Plasmids are engineered in an optimized backbone and are ideal for all transfections.
  • Highest levels of transgene expression in mammalian cells and tissues
  • Suitable for all transfection applications (in vivo and in vitro)
  • High copy number production in Escherichia coli
  • LPS-endotoxin free, supercoiled and highly purified: transfection grade approved
OZ Biosciences proposes five expressions vectors encoding for the most popular reporter genes (CAT, GFP, LacZ, Luciferase, SEAP).
Delivery information: Shipped at room temperature
Packaging: Store at -20 °C