G418 disulfate, Molecular Biology Grade

Supplier: OZ Biosciences

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate

GS21000
103258-196EA 106.68 USD
103258-196
G418 disulfate, Molecular Biology Grade
G418 disulphate

OZ Biosciences G418 Sulfate is an aminoglycoside antibiotic identical to gentamicin B1 produced by Micromonospora rhodorangea


  • Chemical Name: (2R,3S,4R,5R,6S)-5-amino-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl4 methylaminooxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-2-(1-hydroxyethyl)oxane-3,4-diol
  • Molecular Formula: C20H40N4O10. 2 H2SO4
  • Molecular weight: 692.71 g/mol
  • CAS Number: 108321-42-2
  • Molecular biology grade and premium pure


It blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. It is used to select and maintain eukaryotic cells expressing the neo gene (neomycin). The quality and purity of the G-418 is essential to achieve good and consistent selection.


Delivery information: Shipped at room temperature


Packaging: Store at -20°C

Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR