Potassium hydroxide 0.5 M ethanol

Supplier: AQUA SOLUTIONS MS
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

7261-1L
103193-928EA 168 USD
103193-928
Potassium hydroxide 0.5 M ethanol
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1993
ADR: 3 Ⅱ
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR