Dihydroartemisinin

Supplier: 0000027161

Synonyms: α-Dihydroartemisinin

AG-CN2-0468-M050 AG-CN2-0468-M250 AG-CN2-0468-G001
102988-380EA 62.31 USD
102988-380 102988-382 102988-384
Dihydroartemisinin
Dihydroartemisinin

Active metabolite of artemisinin. Anti-malaria (Plasmodium). Anti-schistosoma. Apoptosis inducer. Anti-cancer. Anti-inflammatory Inhibits production of tumor necrosis factor-alpha (TNF-alpha), interleukin-6 (IL-6), monocyte chemotactic protein-1 (MCP-1) and nitric oxide (NO). Autophagy inducer.

Formula: C₁₅H₂₄O₅
MDL Number: MFCD00274495
CAS Number: 71939-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR