Hydrocortisone Hemisuccinate, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LS-1007
102970-982EA 72 USD
102970-982
Hydrocortisone Hemisuccinate, Lifeline Cell Technology
Cell Culture Supplements and Additives
Hydrocortisone Hemisuccinate is used with fibroblasts, basal medium, and endothelial cells.
  • Used with human fibroblasts
  • Used with human endothelial cells
  • Used with basal medium
Hydrocortisone Hemisuccinate LifeFactor may be used with: FibroLife®basal medium and human fibroblasts, VascuLife® basal medium and human endothelial cells.