DL-Pantolactone ≥97%

Supplier: Matrix Scientific

Synonyms: Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, DL-Pantoyl Lactone, 3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, DL-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone

085303-1G 085303-10G 085303-50G
102844-700EA 44.62 USD
102844-700 102844-702 102844-704
DL-Pantolactone ≥97%
DL-Pantolactone

Purity: 97% CAS: 79-50-5 MDL Number: MFCD00064333 Molecular Formula: C6H10O3 Molecular Weight: 130,14 Hazard: Irritant

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00064333
CAS Number: 79-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR