2-Thien-2-ylisonicotinonitrile ≥97%

Supplier: Matrix Scientific

085113-1G
102844-252EA 553.85 USD
102844-252
2-Thien-2-ylisonicotinonitrile ≥97%
2-Thien-2-ylisonicotinonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂S
MDL Number: MFCD13195625
CAS Number: 501378-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR