Omniprobe 4 Post, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75964-10
102773-590PK 201.77 USD
102773-590
Omniprobe 4 Post, Electron Microscopy Sciences
Graticules, Reticles and Grids
The 4 post has copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with 'V'shaped alighment surfaces. Sides have lower profile for easier access to outermost posts.
Packaging: Packaged in plastic vials.