Denka M3 LaB6 Cathode, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80904 80912 80900 80908 80902 80906 80910 80914 80900-10 80900-100 80900-101 80900-11 80900-20 80900-21 80900-40 80900-41 80900-5 80900-60 80900-61 80900-80 80904-10 80904-100 80904-101 80904-20 80904-21 80904-40 80904-41 80904-5 80904-60 80904-61 80904-80 80904-81 80906-10 80906-100 80906-11 80906-20 80906-21 80906-40 80906-41 80906-5 80906-60 80906-61 80906-80 80906-81 80908-10 80908-100 80908-11 80908-20 80908-21 80908-40 80908-41 80908-5 80908-60 80908-61 80908-80 80908-81 80914-20
100492-392EA 2377.89 USD
100492-392 100502-150 100502-144 100502-146 100492-390 100492-394 100502-148 102680-468 102681-392 102681-394 102681-396 102681-398 102681-400 102681-402 102681-404 102681-406 102681-408 102681-410 102681-412 102681-414 102681-416 102681-418 102681-420 102681-422 102681-424 102681-426 102681-428 102681-430 102681-432 102681-434 102681-436 102681-438 102681-440 102681-442 102681-444 102681-446 102681-448 102681-450 102681-452 102681-454 102681-456 102681-458 102681-460 102681-462 102681-464 102681-466 102681-468 102681-470 102681-472 102681-474 102681-476 102681-478 102681-480 102681-482 102681-484 102681-486 102681-488
Denka M3 LaB6 Cathode, Electron Microscopy Sciences
Lights Hollow Cathode Lamps
Lanthanum Hexaboride is the optimum cathode material for use in SEM, TEM, X-Ray Microanalysis, and Electron Beam Lithography.
  • Low potential barrier (work function)
  • High melting point and stability at high temperatures
  • Low vapor pressure-reduces evaporation lost
  • Chemical stability
LaB6 beam is ten times brighter than tungsten, has superior resolution-wide range of acceleration voltages. It provides sharp and clear pictures even to the smallest details. The initial brightness of a Denka Model 3 cathode (1×106 A/cm2.Str) is substantially better than thatMore Product Information