2,2,6,6-Tetramethylpiperidine 99, Liquid

Supplier: CHEM IMPEX INTL INC MS

24504-25G 24504-1KG 24504-250G 24504-100G 24504-100G 24504-250G 24504-25G
102601-562EA 27.08 USD
102601-562 102601-564 102613-650 102625-146 CH24504-100G CH24504-250G CH24504-25G
2,2,6,6-Tetramethylpiperidine 99, Liquid
2,2,6,6-Tetramethylpiperidine

2,2,6,6-Tetramethylpiperidine can be used as a hindered base (hindered amine) because it can dissolve in organic solvents unlike inorganic bases such as potassium hydroxide.


Synonym: Norpempidine

Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152
Density: 0.837 g/mL (25 °C)
Flash Pt: 24
Storage Temperature: Freezer
MDL Number: MFCD00005985
CAS Number: 768-66-1
UN: 1992
ADR: 3 III

Order Now

Specification Test Results

Refractive Index (n20/D) 1.445
Purity (GC) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR