3-Amino-1H-pyrazole

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

24487-5G
102625-126EA 27.08 USD
102625-126
3-Amino-1H-pyrazole
3-Amino-1H-pyrazole
Formula: C₃H₅N₃
MDL Number: MFCD00005236
CAS Number: 1820-80-0
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR