Acridine orange (hemizinc salt), redish brown powder biological stain

Supplier: CHEM IMPEX INTL INC MS

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

00481-5G 00481-25G 00481-100G 00481-250G 00481-5G
102615-288EA 29.23 USD
102615-288 102615-290 102615-292 102615-294 CH00481-5G
Acridine orange (hemizinc salt), redish brown powder biological stain
Acridine orange (hemizinc salt)
Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS Number: 10127-02-3

Order Now

Specification Test Results

Loss on Drying ≤1%
Dye Content ≥90.0%
Absorption at Lambda Max at 492 nm ≥0.514 (5.0 mg/l in Water)
Absorption at Lambda Max at 269 nm ≥0.514 (5.0 mg/l in Water)
Extinction Coefficient at 489 - 495 nm ≥38000
Extinction Coefficient at 265 - 271 nm ≥38000
Solubility Clear, orange to brown solution (1 mg/mL in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR