1-Methylindole ≥98%, light yellow, clear liquid

Supplier: CHEM IMPEX INTL INC MS

Synonyms: N-Methylindole, 1-Methyl-1H-indole

22008-25G 22008-100G 22008-250G 22008-1KG 22008-100G
102568-650EA 36.74 USD
102568-650 102580-970 102580-972 102609-160 CH22008-100G
1-Methylindole ≥98%, light yellow, clear liquid
1-Methylindole
Formula: C₉H₉N
Boiling Pt: 236…239 °C
Density: 1.05
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005800
CAS Number: 603-76-9
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Purity by HPLC ≥98%
Specific Gravity 1.04-1.05 g/mL
Appearance Light yellow liquid
Refractive Index 1.606 - 1.608
Storage Store at 0-8 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR