4-Bromoanisole ≥95%

Supplier: Matrix Scientific

075951-10G 075951-100G
102543-778EA 61.54 USD
102543-778 102543-780
4-Bromoanisole ≥95%
4-Bromoanisole

Hazard: Irritant

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 223
Melting Pt: 10
Density: 1.494
Flash Pt: 94 °C
MDL Number: MFCD00000097
CAS Number: 104-92-7
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR