1-Bromo-4-ethynylbenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne

074723-5G
102537-882EA 176.92 USD
102537-882
1-Bromo-4-ethynylbenzene ≥95%
1-Bromo-4-ethynylbenzene

Hazard: Irritant

Formula: C₈H₅Br
MW: 181.03 g/mol
MDL Number: MFCD00168822
CAS Number: 766-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR