5-Methyl-4-nitro-3-isoxazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

073002-250MG 073002-1G 073002-5G
102527-866EA 498.46 USD
102527-866 102527-868 102527-870
5-Methyl-4-nitro-3-isoxazolecarboxylic acid ≥95%
5-Methyl-4-nitro-3-isoxazolecarboxylic acid

Hazard: Irritant

Formula: C5H4N2O5
MDL Number: MFCD11520861
CAS Number: 960225-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR