3-Amino-2,6-dibromopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dibromopyridin-3-amine

072844-5G
102527-050EA 23.08 USD
102527-050
3-Amino-2,6-dibromopyridine ≥95%
3-Amino-2,6-dibromopyridine

Hazard: Irritant

Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD00128862
CAS Number: 39856-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR