LaboPlast® Compact Jerrycan, Burkle
Supplier: BURKLE INC
0435-0001 0435-1050 0435-2050 0435-3000 0435-1100 0435-2100 0435-0010 0435-3010
10216-896EA 131.44 USD
10216-896 10216-898 10216-900 10216-902 76183-096 76183-098 76183-100 76183-156
LaboPlast® Compact Jerrycan, Burkle
Bottles Jerricans
The compact Jerrycan is ideal as a space-saving canister, with available widths of 6.5 cm (2.56") or 12.5 cm (4.91").
  • Available with or without ³/₄" threaded connector
  • Polypropylene, transparent
  • Includes tamper-proof closure
  • Canister without stopcock is autoclavable
  • Equipped with moulded, indestructible liter graduations
Different liquids can be stored and decanted in a small space.
Compact Jerrycan and Stopcock Set
The compact jerrycan and stopcock set consists of the compact jerrycan with glands, in 5 L (1.32 gal.) or 10 L (1.64 gal.) sizes, andMore Product Information