Palladium, annular sputtering target, Diameter 87×57 mm (OD×ID)

Supplier: Electron Microscopy Sciences

91031-PD
102099-832EA 1540.72 USD
102099-832
Palladium, annular sputtering target, Diameter 87×57 mm (OD×ID)
Palladium

Metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.

Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR