Iron, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-FE 91007-FE 91008-FE 91009-FE 91010-FE 91013-FE 91014-FE 91015-FE 91016-FE 91017-FE 91017-IR 91018-FE 91019-FE 91020-FE
102099-404EA 74.74 USD
102099-404 102099-434 102099-464 102099-494 102099-522 102099-554 102099-584 102099-614 102099-644 102099-674 102099-676 102099-704 102099-734 102099-764
Iron, sputtering target sputter coater cathodes
Iron

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
MDL Number: MFCD00010999
CAS Number: 7439-89-6
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR