Palladium, sputtering target sputter coater cathodes

Supplier: Electron Microscopy Sciences

91006-PD 91007-PD 91008-PD 91009-PD 91010-PD 91013-PD 91014-PD 91015-PD 91016-PD 91017-PD 91018-PD 91019-PD 91020-PD
102099-412EA 808.58 USD
102099-412 102099-442 102099-472 102099-502 102099-530 102099-562 102099-592 102099-622 102099-652 102099-682 102099-712 102099-742 102099-772
Palladium, sputtering target sputter coater cathodes
Palladium

EMS (ELECTRON MICROSCOPY SCIENCES) precious metal sputtering targets are made from high purity metals, starting 99.99% with most standard disc sizes, for use on most sputtering units from many manufacturers: Emitech, Emscope, Bio-Rad, Polaron, Edwards, Balzers, Plasma Sciences, Technics Hummers, Denton, Cressington, and much more.


EMS targets come in a standard thickness of 0.1 mm (100µm). Other thickness, ranging from 0.05 to 6mm, are available upon request. Targets are avaible mounted or unmounted whichever you prefer. For mounting please always state the make and the model of the sputtering unit so we can tend to it for you. Targets in other materials such as Chromium, Iridium etc. are available upon request. We also can manufacture targets with diameters from 4 mm to 304.8 mm (12"). Call for pricing.

Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR