Secure-Seal™ Hybridization Chambers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70333-34 70333-42 70333-44 70333-30 70333-10 70333-32 70333-36 70333-46 70333-20 70333-40 70333-22 70333-50 70333-01 70333-02 70333-03 70333-04 70333-27 70333-37 70333-47 70333-58 70333-62 70333-66 70333-70 70333-74 70333-76 70333-82 70333-83 70333-86
100489-050PK 353.11 USD
100489-050 100489-056 100489-058 100501-878 100489-042 100489-048 100489-052 100501-208 100489-044 100489-054 100489-046 100489-060 102096-662 102096-664 102096-666 102096-668 102096-670 102096-672 102096-674 102096-676 102096-678 102096-680 102096-682 102096-684 102096-686 102096-688 102096-690 102096-692
Secure-Seal™ Hybridization Chambers, Electron Microscopy Sciences
Slides Microarray Slides
Design minimize friction, promotes reagent mixing, and facilitates uniform hybridization.
  • Sealable access ports (1.5 mm) in the chamber surface allow for the addition and removal of reactants
  • Adhesive seal tabs create leak-proof chambers that are temperature resistant
  • SecureSeal™adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating
  • SecureSeals™are ideally suited for protocols utilizing autoradiographic, fluorescent or cheminescent end points
  • Polycarbonate HybriSlip used, 0.25 mm thick
SecureSeal™ adhesiveMore Product Information