Molybdenum, disc, drilled apertures 6.35 mm O.D., Thickness 0.006 mm

Supplier: Electron Microscopy Sciences

47030-MO 47050-MO 47100-MO 47150-MO 47200-MO
102092-694EA 421.77 USD
102092-694 102092-696 102092-702 102092-704 102092-706
Molybdenum, disc, drilled apertures 6.35 mm O.D., Thickness 0.006 mm
Molybdenum

Hole Sizes not listed are available upon special request.

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
MDL Number: MFCD00003465
CAS Number: 7439-98-7
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR