Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm

Supplier: Electron Microscopy Sciences

43010-MO 43020-MO 43030-MO 43050-MO 43060-MO 43070-MO 43100-MO 43150-MO 43200-MO 43300-MO
102092-622EA 242.75 USD
102092-622 102092-624 102092-626 102092-628 102092-630 102092-632 102092-634 102092-636 102092-638 102092-640
Molybdenum, disc, drilled apertures 3 mm O.D., Thickness 0.12 mm
Molybdenum

Hole Sizes not listed are available upon special request.

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
MDL Number: MFCD00003465
CAS Number: 7439-98-7
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR