Carbon Rods, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70210 70212 70230 70200 70231 70301 70302
100488-372PK 182.43 USD
100488-372 100488-374 100501-170 100488-370 102091-132 102091-134 102091-136
Carbon Rods, Electron Microscopy Sciences
Solid Rods
Carbon /graphite films are ideal in EM due to uniform amorphous form and high transparency to electron beams. Carbon/graphite rods are CVP™ (Chemical Vapor Process) purity and spectrographic grade, grade 1, produced to ASTM tolerances. Spectro-Grade rods are ideal for all TEM, SEM and all other critical applications.
  • Spectro-Grade graphite rods have an impurity level of 2 ppm or less with a maximum level element of each of 1 ppm
  • Technical grade carbon has a typical impurity level of 50 ppm or greater
  • For non critical SEM coating applications, technical
More Product Information