Biotube™ Cluster Tube System, Simport
Supplier: Simport Scientific
T100-2B T100-4R T100-20 T100-40 T100-26 T100-1B T100-1G T100-1R T100-2G T100-2Y T100-3R T100-3Y T100-3B T100-4B T100-4G T100-35 T100-28 T100-3G T100-1Y T100-2R T100-4Y T100-25 T100-30 T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-50B T100-50G T100-50R T100-20LST
60872-692CS 156.53 USD
60872-692 60872-712 60872-716 60870-028 60870-022 60872-684 60872-686 60872-688 60872-694 60872-698 60872-700 60872-702 60872-706 60872-708 60872-710 60872-722 60870-026 60872-704 60872-690 60872-696 60872-714 60872-718 60872-720 10199-456 10199-544 10199-546 10199-548 10199-550 10199-558 10199-630 10199-632 10199-640
Biotube™ Cluster Tube System, Simport
Tubes Microtubes
The Biotube™ Cluster Tube System is a polypropylene tube and rack system with tubes held in a standard 8×12 configuration.
  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Ideal for storing, freezing and transporting reagents and specimens
  • Racks and tubes can be used over a wide temperature range, −80 to 121 °C
Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as aMore Product Information