Quartz Microfiber Filter Paper, Grade MK 360, Ahlstrom
Supplier: Ahlstrom
3600-0470
10199-564PK 106.27 USD
10199-564
Quartz Microfiber Filter Paper, Grade MK 360, Ahlstrom
Filters Glass Fiber Filters
Micro-quartz filters can be used in the analysis of hot 'stack' gases and other acidic gases (excluding HF).
They contain minimal traces of metals and minerals with each batch trace element certified. Temperature resistant up to 900 °C.