Doyen Jansen Mouth Gag, Adult, OR Grade, Sklar®
Supplier: Sklar
75-1025
10198-628EA 924.14 USD
10198-628
Doyen Jansen Mouth Gag, Adult, OR Grade, Sklar®
Spatulas
Sklar's® Doyen-Jansen adult mouth gag, which may also be known as a Molt mouth gag, is used to retain the mouth opening during oral and ENT procedures.
It is a finger ring instrument that features a Tru-grip cam ratchet locking system.