Push Caps for Tubes, Nova Biostorage
Supplier: NBS SCIENTIFIC, LLC MS
MP53001 MP53004PK MP53004 MP53007 MP53009 MP53041 MP53065 MP53068 MP53073 MP53072 MP53043 MP53044 MP53069 MP53077 MP53066 MP53045 MP53067 MP53046 MP53071
101975-670CS 2214 USD
101975-670 101975-672 101975-674 101975-686 101975-694 101975-714 103258-652 103258-658 103258-660 103258-662 103258-664 103258-666 103258-668 103258-670 103258-672 103258-674 103258-676 103258-678 103258-680
Push Caps for Tubes, Nova Biostorage
Closures Test/Sample Tube Closures
These micronic TPE multi pierceable capcluster are used to seal 96 individual tubes.
  • Sterile, gamma radiated (15 kGy) or EtO treated
  • Pre-capped on micronic tubes
  • Split septum to pierce with a pipette tip
  • Resists high chemical solvents
  • Temperature range: –80 to +100 °C
  • Easy visual differentiation with different colors
The Capcluster-96 is a sheet of 96 push caps that can be used to seal an entire rack of 96 tubes at once. After the capcluster is used to seal a rack of push cap tubes, the foil backing can be removed to leave each tube individuallyMore Product Information