Cytidine, [5-3H(N)] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT615-250UCI MT615-1MCI MT615-5MCI
101973-210EA 2700.38 USD
101973-210 101973-212 101973-214
Cytidine, [5-3H(N)] ≥97%, MORpure™
Cytidine, [5-3H(N)]
CAS Number: 14419-78-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR