Sucrose, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1956-50UCI
101972-866EA 1601.92 USD
101972-866
Sucrose, [3H] ≥97%, MORpure™
Sucrose, [3H]
CAS Number: 112642-98-5
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR