Z-D,L-Serine, [3-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC2344-50UCI
101971-456EA 5387.69 USD
101971-456
Z-D,L-Serine, [3-14C] ≥97%
Z-D,L-Serine, [3-14C]
CAS Number: 2088204-21-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR