5-Fluorouracil, [2-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC101-50UCI MC101-250UCI
101970-058EA 5387.69 USD
101970-058 101970-060
5-Fluorouracil, [2-14C] ≥98%, MORpure™
5-Fluorouracil, [2-14C]
CAS Number: 766-63-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR