Lebsche Sternum Chisel, OR Grade, Sklar
Supplier: Sklar
55-2010
10197-516EA 758.24 USD
10197-516
Lebsche Sternum Chisel, OR Grade, Sklar
Chisels
Sklar's Lebsche Sternum Chisel is used in procedures that require splitting of the sternum.
Often a mallet is used in conjunction with the chisel so that more force can be applied in the process. Splitting the sternum may be required in planned cardiovascular or thoracic procedures, or in emergency situations.