Muldoon Lacrimal Dilator, OR Grade, Sklar
Supplier: Sklar
65-6952
10197-420EA 69.08 USD
10197-420
Muldoon Lacrimal Dilator, OR Grade, Sklar
Dilators
Muldoon Lacrimal Dilators are constructed of stainless steel.
  • Non sterile, latex free and reusable
This fine pattern instrument is designed with a knurled handle.