7-Deazaadenosine ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-7-(β-D-ribofuranosyl)pyrrolo[2,3-d]pyrimidine

M1808-1MG M1808-5MG
101969-672EA 294.23 USD
101969-672 101969-674
7-Deazaadenosine ≥98%, MORpure™
7-Deazaadenosine
Formula: C₁₁H₁₄N₄O₄
MDL Number: MFCD00056012
CAS Number: 69-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR