Lambotte Osteotomes, OR Grade, Sklar
Supplier: Sklar
40-6532 40-6534 40-6536 40-6538 40-6540 40-6542 40-6545 40-6548 40-6549 40-6550 40-6552 40-6560 40-6561 40-6562 40-6563 40-6564 40-6565 40-6567 40-6570 40-6571 40-6572 40-6573 40-6574 40-6575 40-6577 40-6535 40-6537
10195-990EA 118.64 USD
10195-990 10195-992 10195-994 10195-996 10195-998 10196-000 10196-002 10196-004 10196-006 10196-008 10196-010 10196-012 10196-014 10196-016 10196-018 10196-020 10196-022 10196-024 10196-026 10196-028 10196-030 10196-032 10196-034 10196-036 10196-038 75817-940 75817-942
Lambotte Osteotomes, OR Grade, Sklar
Chisels
Sklar's® Lambotte osteotomes are desigend with a delicate pattern for precise cutting of bones. The size is best used for small delicate bones.
  • Industry standard
  • Sharp tip configuration
The product is available in straight and curved with a variety of blade widths.