4-Bromopyridinium chloride ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

069611-100G
101941-078EA 146.15 USD
101941-078
4-Bromopyridinium chloride ≥98% for synthesis
4-Bromopyridinium chloride

Hazard code(s): Irritant, Hygroscopic

Formula: C₅H₅BrClN
Melting Pt: ∼270 °C (dec.)
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR