Beeson Plaster Spreader, OR Grade, Sklar
Supplier: Sklar
11-1812
10194-980EA 906.44 USD
10194-980
Beeson Plaster Spreader, OR Grade, Sklar
Retractors
Sklar's® Beeson Plaster Spreader is used to aid in the removal of plaster casts. It has a solid straight handle and two large, flat, rectangular blades with horizontal serrations on the outside of the blade.
  • Once a cut is made in the plaster cast, the blades of the spreader are closed by opening the handle and are inserted to a section of the cast
By squeezing the handle, the blades spread open, there by spreading apart sections of the cast. After multiple sections of the cast are spread open, the cast may be removed.