Keyes Skin Biopsy Set On Base, OR Grade, Sklar
Supplier: Sklar
06-4100
10194-884PK 721.27 USD
10194-884
Keyes Skin Biopsy Set On Base, OR Grade, Sklar
Biopsy Punches
The Sklar Keyes Biopsy Punch is used to remove skin samples for biopsy
This set includes a universal handle with 6 interchangeable punch tips ranging from 2mm to 7mm arranged on a sterilizable storage base. The Keyes punches are often used for gynecological biopsies.