6-Bromo-2-naphthoic acid ≥95%

Supplier: Matrix Scientific

068817-5G
101937-804EA 23.08 USD
101937-804
6-Bromo-2-naphthoic acid ≥95%
6-Bromo-2-naphthoic acid

Hazard: Irritant

Formula: C₁₁H₇BrO₂
MDL Number: MFCD01075720
CAS Number: 5773-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR