2-Aminothiazole-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 2-Aminothiazole-5-carboxaldehyde, 2-Amino-1,3-thiazole-5-carboxaldehyde

038933-500MG
101846-492EA 387.69 USD
101846-492
2-Aminothiazole-5-carbaldehyde
2-Aminothiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₄H₄N₂OS
MDL Number: MFCD02179570
CAS Number: 1003-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR