Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5140 91-5142
10184-458EA 1391.4 USD
10184-458 10184-460
Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomy or dilation and curettage.
  • It is weighted with a medium, 21/2 lbs.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleanedMore Product Information