2,1,3-Benzothiadiazol-4-ylamine

Supplier: Matrix Scientific

Synonyms: 4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole

027723-500MG
101824-742EA 290.77 USD
101824-742
2,1,3-Benzothiadiazol-4-ylamine
2,1,3-Benzothiadiazol-4-ylamine

Hazard: Irritant

Formula: C₆H₅N₃S
MDL Number: MFCD00005810
CAS Number: 767-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR