N-(5-Amino-2-fluorophenyl)-4-(pentyloxy)benzamide

Supplier: Matrix Scientific

025685-500MG
101820-748EA 306.15 USD
101820-748
N-(5-Amino-2-fluorophenyl)-4-(pentyloxy)benzamide
N-(5-Amino-2-fluorophenyl)-4-(pentyloxy)benzamide

CAS: 1020057-03-7 MDL Number: MFCD09997401 Molecular Formula: C18H21FN2O2 Molecular Weight: 316,38 Hazard: Irritant

Formula: C₁₈H₂₁FN₂O₂
MDL Number: MFCD09997401
CAS Number: 1020057-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR