5,6-Dihydroxy-1,3-benzodioxole ≥98%

Supplier: Matrix Scientific

016658-100MG
101805-156EA 429.23 USD
101805-156
5,6-Dihydroxy-1,3-benzodioxole ≥98%
5,6-Dihydroxy-1,3-benzodioxole

Hazard: Irritant

Formula: C₇H₆O₄
MDL Number: MFCD06656537
CAS Number: 21505-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR