Tetrakis(triphenylphosphine)palladium(0) tech, tech. for synthesis

Supplier: Matrix Scientific

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

006395-5G
101790-502EA 207.69 USD
101790-502
Tetrakis(triphenylphosphine)palladium(0) tech, tech. for synthesis
Tetrakis(triphenylphosphine)palladium (0)

Hazard code(s): Moisture Sensitive, Store in Freezer, Stored under Argon

Formula: C₇₂H₆₀P₄Pd
Melting Pt: 103…107 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now

Specification Test Results

Melting point 100-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR