Epredia™ Blue Nylon Biopsy Bags, Epredia
Supplier: Epredia
6774015 6774009 6774010 6774011 6774012 6774013 6774014
89212-290CS 538.74 USD
89212-290 10170-218 10170-220 10171-076 10171-078 10171-080 10171-082
Epredia™ Blue Nylon Biopsy Bags, Epredia
Bags Sample Bags
Ensure unrestricted fluid movement around tissue with Epredia™ Blue Nylon biopsy bags.
  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag