Kimtech™ A5 Cleanroom Coveralls, Nonsterile, Bulk Packed, Kimberly-Clark
Supplier: Ansell Healthcare
49831 49832 49833 49834 49835 49836 49837 49838 49841
10159-978CS 450.87 USD
10159-978 10159-980 10160-670 10160-672 10160-674 10160-676 10160-678 10160-680 10160-682
Kimtech™ A5 Cleanroom Coveralls, Nonsterile, Bulk Packed, Kimberly-Clark
Disposable Garments
High-value combination of quality and comfort, as well as reliable cleanroom protection, trust Kimtech™ A5 Nonsterile cleanroom coveralls.
  • Breathable SMS fabric
  • Clean processed
  • High-neck collar for added protection and reduced contamination risk
  • Zipper flap to cover the zipper area
  • Thumb loops for added protection at the wrist, and to keep sleeves from rolling back
  • Bound, tightly sewn seams offer added protection and strengthen barriers
  • Bulk packaged for convenience and waste reduction
  • Recyclable through the RightCycle™ by Kimberly-Clark Professional®
More Product Information