VWR® Advanced Dura-Shaker for Extreme Environments
Supplier: VWR International
10159-960 10159-964
10159-960EA 5875.23 USD
10159-960 10159-964
VWR® Advanced Dura-Shaker for Extreme Environments
Shakers and Mixers Orbital Shakers
The VWR® Dura-Shakers are designed for a wide range of applications including cell cultures that require CO₂ and humidity for optimal cell growth. The remote control module is designed to sit outside of the incubator. The thin ribbon cable is 5.5 feet (168 cm) long and easily passes underneath incubator door or through incubators utility port (minimum diameter 1.2” (3 cm). Controller magnetically attaches to most incubator doors or can sit on a lab bench.
  • Dura-shaker is designed for use in CO2 incubators and other extreme environments up to 100% humidity
More Product Information